Assay Details
Gene Symbol: MBOAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075152.1 | NP_001068620.1 | 7 - 8 | 1286 | 68 | |
GenBank mRNA | BC119932.1 | - | 7 - 8 | 1286 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
