Assay Details
Gene Symbol: CIPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075242.1 | NP_001068710.1 | 3 - 4 | 407 | 84 | |
GenBank mRNA | BC123445.1 | - | 3 - 4 | 407 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
