Assay Details
Gene Symbol: METTL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075246.2 | NP_001068714.1 | 5 - 6 | 685 | 96 | |
GenBank mRNA | BC120275.1 | - | 5 - 6 | 687 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
