Assay Details
Gene Symbol: RAB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075499.1 | NP_001068967.1 | 6 - 7 | 492 | 76 | |
GenBank mRNA | BC122650.1 | - | 6 - 7 | 492 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
