Assay Details
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075506.1 | NP_001068974.1 | 4 - 5 | 1965 | 125 | |
GenBank mRNA | BC120039.1 | - | 4 - 5 | 1965 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
