Assay Details
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075506.1 | NP_001068974.1 | 5 - 6 | 2128 | 110 | |
GenBank mRNA | BC120039.1 | - | 5 - 6 | 2128 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
