Assay Details
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075506.1 | NP_001068974.1 | 6 - 7 | 2238 | 81 | |
GenBank mRNA | BC120039.1 | - | 6 - 7 | 2238 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
