Assay Details
Gene Symbol: RGS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075596.1 | NP_001069064.1 | 1 | 464 | 116 | |
GenBank mRNA | BC120026.1 | - | 1 | 464 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
