Assay Details
Gene Symbol: RBM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075636.1 | NP_001069104.1 | 11 - 12 | 1353 | 61 | |
GenBank mRNA | BC122593.1 | - | 11 - 12 | 1353 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
