Assay Details
Gene Symbol: RBM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075636.1 | NP_001069104.1 | 12 - 13 | 1488 | 83 | |
GenBank mRNA | BC122593.1 | - | 12 - 13 | 1488 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
