Assay Details
Gene Symbol: SYPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075676.1 | NP_001069144.1 | 4 - 5 | 624 | 105 | |
GenBank mRNA | BC120434.1 | - | 4 - 5 | 624 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
