Assay Details
Gene Symbol: RAP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075700.1 | NP_001069168.1 | 1 - 2 | 357 | 143 | |
GenBank mRNA | BC123730.1 | - | 1 - 2 | 357 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
