Assay Details
Gene Symbol: MYL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075713.1 | NP_001069181.1 | 6 - 7 | 652 | 83 | |
GenBank mRNA | BC118334.1 | - | 6 - 7 | 652 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
