Assay Details
Gene Symbol: STRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075730.1 | NP_001069198.1 | 15 - 16 | 1825 | 71 | |
GenBank mRNA | BC142342.1 | - | 16 - 17 | 1610 | 71 | |
BT026272.1 | - | 15 - 16 | 1825 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
