Assay Details
Gene Symbol: STRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075730.1 | NP_001069198.1 | 4 - 5 | 714 | 84 | |
GenBank mRNA | BC142342.1 | - | 5 - 6 | 499 | 84 | |
BT026272.1 | - | 4 - 5 | 714 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
