Assay Details
Gene Symbol: STRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075730.1 | NP_001069198.1 | 5 - 6 | 729 | 72 | |
GenBank mRNA | BC142342.1 | - | 6 - 7 | 514 | 72 | |
BT026272.1 | - | 5 - 6 | 729 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
