Assay Details
Gene Symbol: PLA2G2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075820.2 | NP_001069288.1 | 2 - 3 | 114 | 85 | |
GenBank mRNA | BC122706.1 | - | 2 - 3 | 115 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC122706.1, NM_001075820.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
