Assay Details
Gene Symbol: NMNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075834.1 | NP_001069302.1 | 3 - 4 | 351 | 109 | |
GenBank mRNA | BC119834.1 | - | 3 - 4 | 351 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
