Assay Details
Gene Symbol: SLC25A41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075841.2 | NP_001069309.1 | 2 - 3 | 428 | 66 | |
GenBank mRNA | BC122815.1 | - | 2 - 3 | 429 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
