Assay Details
Gene Symbol: SLC25A41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075841.2 | NP_001069309.1 | 4 - 5 | 690 | 85 | |
GenBank mRNA | BC122815.1 | - | 4 - 5 | 691 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
