Assay Details
Gene Symbol: ADH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075843.1 | NP_001069311.1 | 7 - 8 | 999 | 138 | |
GenBank mRNA | BC120378.1 | - | 7 - 8 | 999 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
