Assay Details
Gene Symbol: GABRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075844.1 | NP_001069312.1 | 5 - 6 | 779 | 59 | |
GenBank mRNA | BC123418.1 | - | 5 - 6 | 779 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
