Assay Details
Gene Symbol: C1QTNF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075846.2 | NP_001069314.1 | 1 - 2 | 39 | 73 | |
GenBank mRNA | BC122841.1 | - | 1 - 2 | 40 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC122841.1, NM_001075846.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
