Assay Details
Gene Symbol: C1QTNF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075846.2 | NP_001069314.1 | 2 - 3 | 220 | 55 | |
GenBank mRNA | BC122841.1 | - | 2 - 3 | 221 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
