Assay Details
Gene Symbol: TSC22D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075860.2 | NP_001069328.1 | 1 - 2 | 376 | 75 | |
GenBank mRNA | BC122722.1 | - | 1 - 2 | 377 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
