Assay Details
Gene Symbol: IP6K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076010.1 | NP_001069478.1 | 5 - 6 | 1140 | 68 | |
GenBank mRNA | BC122746.1 | - | 5 - 6 | 1140 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
