Assay Details
Gene Symbol: ITPKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076011.1 | NP_001069479.1 | 4 - 5 | 1660 | 93 | |
GenBank mRNA | BC119830.1 | - | 4 - 5 | 1660 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
