Assay Details
Gene Symbol: KRT35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076073.1 | NP_001069541.1 | 4 - 5 | 907 | 96 | |
GenBank mRNA | BC119951.1 | - | 4 - 5 | 907 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
