Assay Details
Gene Symbol: TWSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076089.1 | NP_001069557.1 | 1 - 2 | 120 | 76 | |
GenBank mRNA | BC119840.1 | - | 1 - 2 | 120 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC119840.1, NM_001076089.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
