Assay Details
Gene Symbol: FN3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076182.1 | NP_001069650.1 | 1 - 2 | 242 | 73 | |
GenBank mRNA | BC120409.1 | - | 1 - 2 | 242 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
