Assay Details
Gene Symbol: PIP4K2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076196.1 | NP_001069664.1 | 1 - 2 | 206 | 85 | |
GenBank mRNA | BC120101.1 | - | 1 - 2 | 206 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
