Assay Details
Gene Symbol: ACHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076220.1 | NP_001069688.1 | 1 - 2 | 37 | 133 | |
GenBank mRNA | BC123898.1 | - | 1 - 2 | 37 | 133 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC123898.1, NM_001076220.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
