Assay Details
Gene Symbol: SEC22B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076343.2 | NP_001069811.1 | 5 - 6 | 793 | 82 | |
GenBank mRNA | BC102342.1 | - | 5 - 6 | 794 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
