Assay Details
Gene Symbol: NME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076376.1 | NP_001069844.1 | 4 - 5 | 424 | 117 | |
GenBank mRNA | BC102300.1 | - | 4 - 5 | 424 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
