Assay Details
Gene Symbol: PHYHIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076415.1 | NP_001069883.1 | 3 - 4 | 593 | 76 | |
GenBank mRNA | BC119858.1 | - | 3 - 4 | 623 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
