Assay Details
Gene Symbol: GPANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076468.1 | NP_001069936.1 | 2 - 3 | 435 | 145 | |
GenBank mRNA | BC120141.1 | - | 2 - 3 | 435 | 145 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC120141.1, NM_001076468.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
