Assay Details
Gene Symbol: ST14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076538.1 | NP_001070006.1 | 18 - 19 | 2513 | 99 | |
GenBank mRNA | BC122638.1 | - | 18 - 19 | 2513 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
