Assay Details
Gene Symbol: RWDD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076824.1 | NP_001070292.1 | 1 - 2 | 234 | 97 | |
GenBank mRNA | BC123494.1 | - | 1 - 2 | 234 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC123494.1, NM_001076824.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
