Assay Details
Gene Symbol: TNFRSF21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076911.1 | NP_001070379.1 | 5 - 6 | 2147 | 120 | |
GenBank mRNA | BC123657.1 | - | 5 - 6 | 2147 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
