Assay Details
Gene Symbol: CTSZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077835.1 | NP_001071303.1 | 3 - 4 | 549 | 94 | |
GenBank mRNA | BC122603.1 | - | 3 - 4 | 549 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
