Assay Details
Gene Symbol: DUSP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078017.2 | NP_001071485.1 | 1 | 751 | 108 | |
GenBank mRNA | BC122819.1 | - | 1 | 752 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
