Assay Details
Gene Symbol: MYOZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078039.1 | NP_001071507.1 | 1 - 2 | 231 | 60 | |
GenBank mRNA | BC123727.1 | - | 1 - 2 | 231 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC123727.1, NM_001078039.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
