Assay Details
Gene Symbol: MYOZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078039.1 | NP_001071507.1 | 5 - 6 | 645 | 128 | |
GenBank mRNA | BC123727.1 | - | 5 - 6 | 645 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
