Assay Details
Gene Symbol: MRPS30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079595.1 | NP_001073063.1 | 3 - 4 | 848 | 105 | |
GenBank mRNA | BC122665.1 | - | 3 - 4 | 848 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
