Assay Details
Gene Symbol: FAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079626.2 | NP_001073094.1 | 2 - 3 | 190 | 101 | |
GenBank mRNA | BC122668.1 | - | 2 - 3 | 191 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC122668.1, NM_001079626.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
