Assay Details
Gene Symbol: FAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079626.2 | NP_001073094.1 | 5 - 6 | 607 | 123 | |
GenBank mRNA | BC122668.1 | - | 5 - 6 | 608 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
