Assay Details
Gene Symbol: GGPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079801.1 | NP_001073269.1 | 1 - 2 | 155 | 77 | |
GenBank mRNA | BC120313.1 | - | 1 - 2 | 155 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC120313.1, NM_001079801.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
