Assay Details
Gene Symbol: GGPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079801.1 | NP_001073269.1 | 2 - 3 | 255 | 108 | |
GenBank mRNA | BC120313.1 | - | 2 - 3 | 255 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
