Assay Details
Gene Symbol: RGS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080232.2 | NP_001073701.1 | 2 - 3 | 230 | 142 | |
GenBank mRNA | BC122763.1 | - | 2 - 3 | 231 | 142 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC122763.1, NM_001080232.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
