Assay Details
Gene Symbol: RGS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080232.2 | NP_001073701.1 | 3 - 4 | 387 | 131 | |
GenBank mRNA | BC122763.1 | - | 3 - 4 | 388 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
